Anti-APBB1 Goat Polyclonal Antibody
Catalog # PRSI45-261
Supplier: ProSci Inc.
Specifications
- Antigen name:Amyloid Beta (A4) Precursor Protein-Binding, Family B, Member 1
- Antigen symbol:APBB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:amyloid beta A4 precursor protein-binding|APBB1|FE65|adaptor protein FE65a2|stat-like protein|MGC:9072|member 1|amyloid beta (A4) precursor protein-binding|RIR |family B|member 1 (Fe65)
- Concentration:500 ug/mL
- Immunogen:APBB1 antibody was raised against a 13 amino acid synthetic peptide near the C-Terminus of APBB1.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
APBB1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
APBB1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: APBB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse