Anti-AADAT Goat Polyclonal Antibody
Catalog # PRSI42-742
Supplier: ProSci Inc.
Specifications
- Antigen name:Aminoadipate Aminotransferase
- Antigen symbol:AADAT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:KATII|KAT2|alpha-Aminoadipate Aminotransferase
- Concentration:500 ug/mL
- Immunogen:AADAT antibody was raised against a 14 amino acid synthetic peptide near the internal region of AADAT.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
AADAT antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: AADAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human