Anti-ZEB2 Goat Polyclonal Antibody
PRSI42-432
- Antigen name:Zinc Finger E-box Binding Homeobox 2
- Antigen symbol:ZEB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:HSPC082|Smad-interacting protein 1|SIP1|ZFX1B|KIAA0569|Zinc finger E-box-binding homeobox 2|SMADIP1|ZFHX1B|SIP-1|HRIHFB2411|ZEB2
- Concentration:500 ug/mL
- Immunogen:ZEB2 antibody was raised against a 14 amino acid synthetic peptide near the internal region of ZEB2 (aa 545-558).
- Size:100 µg
- Pk:100 µG
ZEB2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
ZEB2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ZEB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human