Anti-RORC Goat Polyclonal Antibody
Catalog # PRSI42-425
Supplier: ProSci Inc.
Specifications
- Antigen name:Retinoid related orphan receptor gamma
- Antigen symbol:RORC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:RZRG|NR1F3|RORG|RZR-GAMMA|TOR
- Concentration:500 ug/mL
- Immunogen:RORC antibody was raised against a 13 amino acid synthetic peptide near the internal region of RORC (aa 200-212).
- Size:100 µg
- Pk:100 µG
Specifications
About this item
RORC antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
RORC antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: RORC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human