Order Entry
Belgium
ContactUsLinkComponent
Anti-Rubella virus Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-Rubella virus Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # PRSI35-358
Supplier:  ProSci Inc.
undefined
Anti-Rubella virus Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # PRSI35-358
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Rubella virus
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Goat
  • Reactivity:
    Virus
  • Western blot:
    Yes
  • Form:
    Liquid
  • Immunogen:
    Strain HPV78
  • Size:
    1 mL
  • Pk:
    1 mL

Specifications

About this item

Purified IgG fraction of antiserum covalently coupled with high purity Isomer I of fluorescein isothiocyanate. Care is taken to insure complete removal of any free fluorescein from the final product.

Rubella virus antibody is in a phosphate saline buffer (0.01M, pH 7.2) containing BSA (10 mg/ml) as a stabilizer and 0.1% sodium azide preservative.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: Rubella virus
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Goat
Isotype:
Reactivity: Virus