Anti-Enterococcus sp Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # PRSI35-179
Supplier: ProSci Inc.
Specifications
- Antigen name:Enterococcus sp
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- Reactivity:Bacteria
- Western blot:Yes
- Form:Liquid
- Immunogen:Enterococcus faecium
- Size:1 mL
- Pk:1 mL
Specifications
About this item
Purified IgG fraction of antiserum covalently coupled with highly purified preparation of horseradish peroxidase (RZ>3). Care is taken to insure adequate conjugation while preserving the maximum enzyme activity. Free enzyme is absent.
Enterococcus sp. antibody is in a phosphate saline buffer with BSA (10mg/ml) and preserved with 0.002% thimerosal.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: Enterococcus sp
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bacteria