Anti-Aspergillus antigen Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
PRSI35-107
- Antigen name:Aspergillus antigen
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Fungus
- Western blot:Yes
- Form:Liquid
- Immunogen:Soluble extract from A. fumigatus, A. flavus, A. niger and A. terreus
- Size:1 mL
- Pk:1 mL
Purified IgG fraction of antiserum covalently coupled with highly purified preparation of horseradish peroxidase (RZ>3). Care is taken to insure adequate conjugation while preserving the maximum enzyme activity. Free enzyme is absent.
- Aspergillus sp. antibody is in a phosphate saline buffer with BSA (10mg/ml) and preserved with 0.002% thimerosal.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: Aspergillus antigen
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Fungi