Order Entry
Belgium
ContactUsLinkComponent
 
Anti-NPHS1 Mouse Monoclonal Antibody
  PRSI32-200
undefined
Anti-NPHS1 Mouse Monoclonal Antibody
  PRSI32-200

 

  • Antigen name:
    Nephrin
  • Antigen symbol:
    NPHS1
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Antigen synonyms:
    Renal glomerulus-specific cell adhesion receptor|NPHN|CNF|SAPAP1
  • Immunogen:
    Ovalbumin.
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Ovalbumin (OVA) is used in variety of laboratories products which contain OVA as a carrier protein. To raise antibodies against small non-immunogenic molecules,called haptens,it is often necessary to couple them to carrier protein,such as OVA.This antibody is a good reagent that may be used for the development of detection assay in ELISA or western blot to test the remaning OVA for clinical products in Vivo or research products In Vitro.

Purified antibody.

Purified antibody in PBS containing 0.03% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NPHS1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: 0