Anti-SLC7A4 Rabbit Polyclonal Antibody
PRSI29-917
- Antigen name:Solute Carrier Family 7 Member 4
- Antigen symbol:SLC7A4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:HCAT3|VH|CAT4|CAT-4
- Immunogen:A synthetic peptide corresponding a region of human SLC7A4.
- Size:50 µg
- Pk:50 µG
SLC7A4 is involved in the transport of the cationic amino acids (arginine, lysine and ornithine).
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SLC7A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human