Anti-ZNF253 Rabbit Polyclonal Antibody
Catalog # PRSI29-067
Supplier: ProSci Inc.
Specifications
- Antigen name:Zinc Finger Protein 253
- Antigen symbol:ZNF253
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human ZNF253.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
ZNF253 may function as a transcription factor. Seem to have a transcriptional repression activity
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ZNF253
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human