Anti-KIF5A Rabbit Polyclonal Antibody
Catalog # PRSI27-967
Supplier: ProSci Inc.
Specifications
- Antigen name:Kinesin Family Member 5A
- Antigen symbol:KIF5A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:NKHC1|NKHC|Kinesin heavy chain isoform 5A|MY050|SPG10|KIF5|Kinesin heavy chain neuron-specific 1|D12S1889
- Immunogen:A synthetic peptide corresponding a region of human KIF5A.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
KIF5A is a member of the kinesin family of proteins. Members of this family are part of a multisubunit complex that functions as a microtubule motor in intracellular organelle transport. Mutations in this gene cause autosomal dominant spastic paraplegia 10.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: KIF5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human