Order Entry
Belgium
ContactUsLinkComponent
Anti-EMX2 Rabbit Polyclonal Antibody
Anti-EMX2 Rabbit Polyclonal Antibody
  PRSI27-381
undefined
Anti-EMX2 Rabbit Polyclonal Antibody
  PRSI27-381

 

  • Antigen name:
    Empty Spiracles Homeobox 2
  • Antigen symbol:
    EMX2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Dog,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    Homeobox protein EMX2|EMX2|Empty spiracles homolog 2
  • Immunogen:
    A synthetic peptide corresponding a region of human EMX2.
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

The homeodomain transcription factor EMX2 is critical for central nervous system and urogenital development. EMX1 along with EMX2 is related to the 'empty spiracles' gene expressed in the developing Drosophila head.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: EMX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse, Rat