Anti-ELF2 Rabbit Polyclonal Antibody
Catalog # PRSI27-380
Supplier: ProSci Inc.
Specifications
- Antigen symbol:ELF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:NERF-1B|EU32|NERF-2|NERF-1a,b|NERF-1A|NERF
- Immunogen:A synthetic peptide corresponding a region of human ELF2.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
The ELF2 gene encodes a protein that physically interacts with AML1 and mediates opposing effects on AML1-mediated transcription of the B cell-specific blk gene.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ELF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse, Rat