Specifications
- Antigen name:Casein Kinase 2, Alpha 2
- Antigen symbol:CSNK2A2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:CK2A2|CSNK2A1
- Immunogen:A synthetic peptide corresponding a region of human CSNK2A2.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
CSNK2A2 belongs to the protein kinase superfamily, Ser/Thr protein kinase family, CK2 subfamily. It contains 1 protein kinase domain. Casein kinases are operationally defined by their preferential utilization of acidic proteins such as caseins as substrates. The alpha and alpha' chains contain the catalytic site. CSNK2A2 participates in Wnt signaling. It phosphorylates 'Ser-392' of p53/TP53 following UV irradiation.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: CSNK2A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: