Order Entry
Belgium
ContactUsLinkComponent
Anti-NETO2 Rabbit Polyclonal Antibody
Anti-NETO2 Rabbit Polyclonal Antibody
Catalog # PRSI26-221
Supplier:  ProSci Inc.
undefined
Anti-NETO2 Rabbit Polyclonal Antibody
Catalog # PRSI26-221
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Neuropilin (Nrp) And Tolloid (Tll)-Like 2
  • Antigen symbol:
    NETO2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    BTCL2|NEOT2
  • Immunogen:
    A synthetic peptide corresponding a region of human NETO2.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

NETO2 is a predicted transmembrane protein containing two extracellular CUB domains followed by a low-density lipoprotein class A (LDLa) domain. It also has an intracellular FXNPXY-like motif, which has been shown in other proteins to be essential for the internalization of clathrin coated pits during endocytosis.This gene encodes a predicted transmembrane protein containing two extracellular CUB domains followed by a low-density lipoprotein class A (LDLa) domain. It also has an intracellular FXNPXY-like motif, which has been shown in other proteins to be essential for the internalization of clathrin coated pits during endocytosis. Alternatively spliced transcript variants have been observed, but they have not been fully characterized.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NETO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse