Anti-EIF2AK1 Rabbit Polyclonal Antibody
EIF2AK1 acts at the level of translation initiation to downregulate protein synthesis in response to stress. The protein is a kinase that can be inactivated by hemin. Two transcript variants encoding different isoforms have been found for this gene.The HRI gene is localized to 7p22 where its 3' end slightly overlaps the 3' end of the gene JTV1. The two genes are transcribed from opposite strands. Studies in rat and rabbit suggest that the HRI gene product phosphorylates the alpha subunit of eukaryotic initiation factor 2. Its kinase activity is induced by low levels of heme and inhibited by the presence of heme. Sequence Note: The sequence AF181071.1 is a chimeric mRNA clone. Only the heme-regulated initiation factor 2-alpha kinase region was propagated into this RefSeq record.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: EIF2AK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
- Size:50 µg
- Western blot:Yes
- Cat. No.:PRSI25-766
- Clonality:Polyclonal
- Reactivity:Human|Rat|Mouse
- Form:Lyophilized
- Antigen synonyms:PRO1362|HRI|heme-regulated eukaryotic initiation factor eIF-2-alpha kinase|hemin-sensitive initiation factor 2-alpha kinase|KIAA1369|heme-regulated repressor|heme sensitive initiation factor 2a kinase|HCR|heme-controlled repressor|heme-regulated inhibitor|heme regulated initiation factor 2 alpha kinase|heme-regulated initiation factor 2-alpha kinase
- Immunogen:A synthetic peptide corresponding a region of human EIF2AK1.
- Host:Rabbit
- Conjugation:Unconjugated
- Antigen symbol:EIF2AK1
- ELISA:Yes



