Anti-ZNF687 Rabbit Polyclonal Antibody
Catalog # PRSI6861
Supplier: ProSci Inc.
Specifications
- Antigen name:Zinc Finger Protein 687
- Antigen symbol:ZNF687
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:ZNF687 antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human ZNF687.
- Tested applications:E, WB, IF
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
Specifications
About this item
ZNF687 Antibody: The zinc finger protein 687 (ZNF687) was initially identified as a translocation partner gene with RUNX1 in patients with acute myeloid leukemia (AML). Little is known of the function of the ZNF687 protein, but it has been shown to weakly interact with the Ring1/Rnf2 RING finger protein member of the Polycomb group of proteins, suggesting it may be involved in the chromatin-modifying complexes essential for embryonic development and stem cell renewal. Other evidence suggests that ZNF687 may be part of a transcriptional network that also includes ZNF592 and ZMYMD8.
Type:
Antigen: ZNF687
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human