Order Entry
Belgium
ContactUsLinkComponent
 
Anti-B9D1 Rabbit Polyclonal Antibody
Catalog # PRSI6715
Supplier:  ProSci Inc.
undefined
Anti-B9D1 Rabbit Polyclonal Antibody
Catalog # PRSI6715
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    B9 Domain-Containing protein 1
  • Antigen symbol:
    B9D1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Antigen synonyms:
    B9 domain-containing protein 1|B9|B9D1|MKSR1|MKS1-related protein 1|EPPB9|MKS9
  • Storage buffer:
    PBS containing 0.02% sodium azide
  • Immunogen:
    B9D1 antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human B9D1.
  • Tested applications:
    E, WB, ICC, IF
  • Purification:
    Affinity chromatography purified via peptide column
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

B9D1 Antibody: Meckel syndrome (MKS) is an embryonic lethal, autosomal recessive disorder characterized by polycystic kidney disease, central nervous system defects, polydactyly and liver fibrosis. B9D1 is a B9 domain-containing protein, one of several that are involved in ciliogenesis. Alterations in expression of this gene have been found in a family with Meckel syndrome. B9D1, and its related protein B9D2, form a complex with MKS1, disruption of which causes MKS. B9D1 is thought to be required for normal hedgehog signaling, ciliogenesis, and ciliary protein localization.

Type:
Antigen: B9D1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human