Anti-SESTD1 Rabbit Polyclonal Antibody
Catalog # PRSI5877
Supplier: ProSci Inc.
Specifications
- Antigen name:SEC14 And spectrin Domains 1
- Antigen symbol:SESTD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Antigen synonyms:SEC14 domain and spectrin repeat-containing protein 1|SOLO|Huntingtin-interacting protein-like protein
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:SESTD1 antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human SESTD1.
- Tested applications:E, WB, IHC, IF
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
Specifications
About this item
SESTD1 Antibody: SESTD1 was initially identified in mutant zebrafish with defects in the spontaneous contraction and touch response as a novel gene, solo, encoding a protein containing SEC14 and spectrin repeat domains. Other experiments indicated that SESTD1 interacts with the TRPC4 and TRPC5, members of the transient receptor potential channel family, via the TRPC calmodulin- and inositol 1, 4, 5-triphosphate receptor-binding domain and is essential for efficient receptor-mediated activation of TRPC5, suggesting that SESTD1 is a novel regulator of these TRPC proteins.
Type:
Antigen: SESTD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat