Order Entry
Belgium
ContactUsLinkComponent
Anti-RNASEH2A Rabbit Polyclonal Antibody
Anti-RNASEH2A Rabbit Polyclonal Antibody
Catalog # PRSI4979
Supplier:  ProSci Inc.
undefined
Anti-RNASEH2A Rabbit Polyclonal Antibody
Catalog # PRSI4979
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Ribonuclease H2, Subunit A
  • Antigen symbol:
    RNASEH2A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Antigen synonyms:
    RNHIA|AGS4|RNase H2 subunit A|RNHL|Aicardi-Goutieres syndrome 4 protein|Ribonuclease H2 subunit A|JUNB|RNASEHI|RNAse H2A
  • Storage buffer:
    PBS containing 0.02% sodium azide
  • Immunogen:
    RNAse H2A antibody was raised against a 17 amino acid synthetic peptide near the center of human RNAse H2A.
  • Tested applications:
    E, WB, ICC, IF
  • Purification:
    Affinity chromatography purified via peptide column
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

RNAse H2A Antibody: Ribonucleases (RNAses) H are enzymes that hydrolyze the RNA strands of RNA/DNA hybrids. The major role of these enzymes is to remove the RNA strand from the RNA/DNA hybrids that form during DNA replication and repair. RNAse H2 is made up of three subunits; all three are required for RNAse activity. Recent evidence has demonstrated that mutations in RNAse H2A or any of the other subunits result in Aicardi-Goutieres syndrome (AGS), a neurological disorder with similar symptoms to viral brain infections including high levels of IFN-alpha in the cerebral spinal fluid. Similar conditions are observed with mutations in TREX1, a single-stranded DNA exonuclease, suggesting that RNAse H2 and TREX1 may have similar roles, and that mutations in any of these genes lead to an accumulation of intracellular nucleic acids, triggering an inflammatory response through activation of the innate immune system.

Type:
Antigen: RNASEH2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat