Anti-MED4 Rabbit Polyclonal Antibody
Catalog # PRSI4729
Supplier: ProSci Inc.
Specifications
- Antigen name:Mediator Complex Subunit 4
- Antigen symbol:MED4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:MED4 antibody was raised against a 17 amino acid synthetic peptide from near the carboxy terminus of human MED4.
- Tested applications:E, WB, IHC
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
Specifications
About this item
MED4 Antibody: The mediator complex is a multi-protein transcriptional co-activator that is expressed ubiquitously in eukaryotes from yeast to mammals and is required for induction of RNA polymerase II (pol II) transcription by DNA binding transcription factor. One of the proteins in this complex is MED4. This protein has also been shown to be part of the vitamin D receptor interacting complex. MED4 was recently identified as an HIV dependency factor (HDF), suggesting that MED4 may be an important drug target in HIV treatment.
Type:
Antigen: MED4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat