Order Entry
Belgium
Orders LinkContactUsLinkComponent
Calcitriol, Reference Standards

Calcitriol, Reference Standards

Supplier:   USP
Calcitriol, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Caution: Cold Shipment required

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1086301.
  • 1086312.
  • Description:
  • Calcitriol
  • Calcitriol Solution
  • CAS No.:
  • 32222-06-3
  • 32222-06-3
  • Formula:
  • C₂₇H₄₄O₃
  • C₂₇H₄₄O₃
  • UN No.:
  • 2811
  • 1230

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1086301.
    Calcitriol
    Calcitriol
    USP reference standard
    Vial
    10 mg
    Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 3 340,00 €
    3 340,00 €
    1 * 10 mg
       3 340,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1086301.
    Description
    Calcitriol
    CAS No.
    32222-06-3
    Formula
    C₂₇H₄₄O₃
    UN No.
    2811
  • 1086312.
    Calcitriol Solution
    Calcitriol Solution
    USP reference standard
    Ampule
    5 mL
    Log in for availability
    Log in for availability
    1 * 5 mL
    1 * 5 mL - 509,00 €
    509,00 €
    1 * 5 mL
       509,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1086312.
    Description
    Calcitriol Solution
    CAS No.
    32222-06-3
    Formula
    C₂₇H₄₄O₃
    UN No.
    1230
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.