Ziprasidone Related Compounds, Reference Standard
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
5 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1724419.
- 1724420.
- 1724441.
- 1724452.
- 1724474.
- Description:
- Ziprasidone Related Compound A
- Ziprasidone Related Compound B
- Ziprasidone Related Compound C
- Ziprasidone Related Compound D
- Ziprasidone Related Compound F
- Synonym(s):
- 3-(Piperazin-1-yl)benzo[d]isothiazole monohydrochloride
- 5-(2-(4-(Benzo[d]isothiazol-3-yl)piperazin-1-yl)ethyl)-6-chloroindoline-2,3-dione
- 5,5'-Bis(2-(4-(benzo[d]isothiazol-3-yl)piperazin-1-yl)ethyl)-6,6'-dichloro-3-hydroxy-3,3'-biindoline-2,2'-dione
- 3-(Benzo[d]isothiazol-3-yl)-5-(2-(4-(benzo[d]isothiazol-3-yl)piperazin-1-yl)ethyl)-6-chloroindolin-2-one
- Sodium 2-(2-amino-5-{2-[4-(benzo[d]isothiazol-3-yl)piperazin-1-yl]ethyl}-4-chlorophenyl)acetate monohydrate
- CAS No.:
- 87691-88-1
- 1159977-56-6
- 1303996-68-0
- 1159977-04-4
- Formula:
- C₁₁H₁₄ClN₃S
- C₂₁H₁₉ClN₄O₂S
- C₄₂H₄₀Cl₂N₈O₃S₂
- C₂₈H₂₄ClN₅OS₂
- UN No.:
- 3077
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1724419.Ziprasidone Related Compound AZiprasidone Related Compound AUSP reference standardVial10 mgLog in for availability1 * 10 mg1 200,00 €Specifications:
More
Specifications
Cat. No.1724419.DescriptionZiprasidone Related Compound ASynonym(s)3-(Piperazin-1-yl)benzo[d]isothiazole monohydrochlorideCAS No.87691-88-1FormulaC₁₁H₁₄ClN₃SUN No.3077 - 1724420.Ziprasidone Related Compound BZiprasidone Related Compound BUSP reference standardVial10 mgLog in for availability1 * 10 mg1 350,00 €Specifications:
More
Specifications
Cat. No.1724420.DescriptionZiprasidone Related Compound BSynonym(s)5-(2-(4-(Benzo[d]isothiazol-3-yl)piperazin-1-yl)ethyl)-6-chloroindoline-2,3-dioneCAS No.1159977-56-6FormulaC₂₁H₁₉ClN₄O₂SUN No. - 1724441.Ziprasidone Related Compound CZiprasidone Related Compound CUSP reference standardVial10 mgLog in for availability1 * 10 mg1 360,00 €Specifications:
More
Specifications
Cat. No.1724441.DescriptionZiprasidone Related Compound CSynonym(s)5,5'-Bis(2-(4-(benzo[d]isothiazol-3-yl)piperazin-1-yl)ethyl)-6,6'-dichloro-3-hydroxy-3,3'-biindoline-2,2'-dioneCAS No.1303996-68-0FormulaC₄₂H₄₀Cl₂N₈O₃S₂UN No. - 1724452.Ziprasidone Related Compound DZiprasidone Related Compound DUSP reference standardVial10 mgLog in for availability1 * 10 mg1 360,00 €Specifications:
More
Specifications
Cat. No.1724452.DescriptionZiprasidone Related Compound DSynonym(s)3-(Benzo[d]isothiazol-3-yl)-5-(2-(4-(benzo[d]isothiazol-3-yl)piperazin-1-yl)ethyl)-6-chloroindolin-2-oneCAS No.1159977-04-4FormulaC₂₈H₂₄ClN₅OS₂UN No. - 1724474.Ziprasidone Related Compound FZiprasidone Related Compound FUSP reference standardVial15 mgLog in for availability1 * 15 mg1 180,00 €Specifications:
More
Specifications
Cat. No.1724474.DescriptionZiprasidone Related Compound FSynonym(s)Sodium 2-(2-amino-5-{2-[4-(benzo[d]isothiazol-3-yl)piperazin-1-yl]ethyl}-4-chlorophenyl)acetate monohydrateCAS No.FormulaUN No.



