Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tazobactam, Reference Standards

Tazobactam, Reference Standards

Supplier:   USP
Tazobactam, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1643383.
  • 1643394.
  • Description:
  • Tazobactam
  • Tazobactam Related Compound A
  • Synonym(s):
  • (2S,3S)-2-Amino-3-methyl-3-sulFood Ingredientsno-4-(1H-1,2,3-triazol-1-yl)butyric acid
  • CAS No.:
  • 89786-04-9
  • 118175-11-4
  • Formula:
  • C₁₀H₁₂N₄O₅S
  • C₇H₁₂N₄O₄S

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1643383.
    Tazobactam
    Tazobactam
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 416,00 €
    416,00 €
    1 * 200 mg
       416,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1643383.
    Description
    Tazobactam
    Synonym(s)
    CAS No.
    89786-04-9
    Formula
    C₁₀H₁₂N₄O₅S
  • 1643394.
    Tazobactam Related Compound A
    Tazobactam Related Compound A
    USP reference standard
    Vial
    20 mg
    Log in for availability
    Log in for availability
    1 * 20 mg
    1 * 20 mg - 1 240,00 €
    1 240,00 €
    1 * 20 mg
       1 240,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1643394.
    Description
    Tazobactam Related Compound A
    Synonym(s)
    (2S,3S)-2-Amino-3-methyl-3-sulFood Ingredientsno-4-(1H-1,2,3-triazol-1-yl)butyric acid
    CAS No.
    118175-11-4
    Formula
    C₇H₁₂N₄O₄S