Orders LinkContactUsLinkComponent
Rods, AR-Glas®
Rods, AR-Glas®
Supplier:   Schott AG
Rods, AR-Glas®
Supplier:  Schott AG
Certificates
13 Options Available Below
Product Details & Documents
Made of AR®-Glas.
  • Soft glass is acid-resistant
  • Has high thermal expansion for pressure melting and for fusing with platinum or dumet and other alloys
Product Family Options
Product Information
  • Ext.Ø
    Length
    Availability
    Price
  • 3±0,10 mm
    1500 mm
    Log in for availability
    Log in for availability
    1 * 13,5 kg
    1 * 13,5 kg - 183,00 €
    183,00 €
    1 * 13,5 kg
       183,00 €
  • 4±0,15 mm
    1500 mm
    Log in for availability
    Log in for availability
    1 * 14,5 kg
    1 * 14,5 kg - 196,00 €
    196,00 €
    1 * 14,5 kg
       196,00 €
  • 5±0,15 mm
    1500 mm
    Log in for availability
    Log in for availability
    1 * 16 kg
    1 * 16 kg - 216,00 €
    216,00 €
    1 * 16 kg
       216,00 €
  • 6±0,15 mm
    1500 mm
    Log in for availability
    Log in for availability
    1 * 15 kg
    1 * 15 kg - 192,00 €
    192,00 €
    1 * 15 kg
       192,00 €
  • 7±0,20 mm
    1500 mm
    Log in for availability
    Log in for availability
    1 * 15 kg
    1 * 15 kg - 192,00 €
    192,00 €
    1 * 15 kg
       192,00 €
  • 8±0,20 mm
    1500 mm
    Log in for availability
    Log in for availability
    1 * 15 kg
    1 * 15 kg - 192,00 €
    192,00 €
    1 * 15 kg
       192,00 €
  • 9±0,20 mm
    1500 mm
    Log in for availability
    Log in for availability
    1 * 14,3 kg
    1 * 14,3 kg - 184,00 €
    184,00 €
    1 * 14,3 kg
       184,00 €
  • 10±0,25 mm
    1500 mm
    Log in for availability
    Log in for availability
    1 * 14,4 kg
    1 * 14,4 kg - 185,00 €
    185,00 €
    1 * 14,4 kg
       185,00 €
  • 12±0,25 mm
    1500 mm
    Log in for availability
    Log in for availability
    1 * 14 kg
    1 * 14 kg - 180,00 €
    180,00 €
    1 * 14 kg
       180,00 €
  • 14±0,30 mm
    1500 mm
    Log in for availability
    Log in for availability
    1 * 13,8 kg
    1 * 13,8 kg - 177,00 €
    177,00 €
    1 * 13,8 kg
       177,00 €
  • 16±0,35 mm
    1500 mm
    Log in for availability
    Log in for availability
    1 * 15,1 kg
    1 * 15,1 kg - 194,00 €
    194,00 €
    1 * 15,1 kg
       194,00 €
  • 20±0,50 mm
    1500 mm
    Log in for availability
    Log in for availability
    1 * 18,8 kg
    1 * 18,8 kg - 241,00 €
    241,00 €
    1 * 18,8 kg
       241,00 €
  • 25±0,70 mm
    1500 mm
    Log in for availability
    Log in for availability
    1 * 16,6 kg
    1 * 16,6 kg - 321,00 €
    321,00 €
    1 * 16,6 kg
       321,00 €