Orders LinkContactUsLinkComponent
Containment berms, QuickBerm® Plus
Containment berms, QuickBerm® Plus
Certificates
Containment berms, QuickBerm® Plus
Containment berms, QuickBerm® Plus
6 Options Available Below
Product Details & Documents
The QuickBerm Plus® offers superior protection and chemical resistance against the harshest of liquids, including petroleum, oils, greases, and most acids.
  • Single-piece construction - patented Rigid-Lock wall-support system integral to berm provides unobstructed work space
  • Quick and easy setup, no assembly required
  • Withstands long-term UV exposure and extreme temperatures
  • Finished hem edge on top of sidewall prevents fraying and adds strength
A simple push on the walls unlocks the support and lays the wall down for drive-through capability from any angle. Reinforced CriticalCorner™ design with wraparound diagonal radio frequency (RF) welds offer exceptionally strong structural integrity to prevent seam leaks, provide fluid-tight reliability, and extend berm life expectancy.
For added resistance against rocky surfaces or heavy in-out traffic use tarps, ground mats, track mats, or track runners. Excellent for heavy vehicle traffic
 : Helps to comply with EPA for containment and spill prevention (SPCC)
 : Anchors not included.
Product Family Options
Product Information
  • Capacity
    W×D×H
    Availability
    Price
  • 119-0358
    Rigid-lock QuickBerm® plus, folded dimensions (610×610×305 mm), 24 kg
    Rigid-lock QuickBerm® plus, folded dimensions (610×610×305 mm), 24 kg
    2252 l
    3000×2603×305 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 160,00 €
    2 160,00 €
    1 * 1 ST
       2 160,00 €
  • 119-0359
    Rigid-lock QuickBerm® plus, folded dimensions (610×610×305 mm), 37,6 kg
    Rigid-lock QuickBerm® plus, folded dimensions (610×610×305 mm), 37,6 kg
    4240 l
    4600×3000×305 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 090,00 €
    3 090,00 €
    1 * 1 ST
       3 090,00 €
  • 119-0360
    Rigid-lock QuickBerm® plus, folded dimensions (610×610×305 mm), 68,0 kg
    Rigid-lock QuickBerm® plus, folded dimensions (610×610×305 mm), 68,0 kg
    9501 l
    8500×3700×305 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5 300,00 €
    5 300,00 €
    1 * 1 ST
       5 300,00 €
  • 119-0361
    Rigid-lock QuickBerm® plus, folded dimensions (610×610×305 mm), 131,5 kg
    Rigid-lock QuickBerm® plus, folded dimensions (610×610×305 mm), 131,5 kg
    16978 l
    15200×3700×305 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7 290,00 €
    7 290,00 €
    1 * 1 ST
       7 290,00 €
  • 119-0362
    Rigid-lock QuickBerm® plus, folded dimensions (610×610×305 mm), 139,3 kg
    Rigid-lock QuickBerm® plus, folded dimensions (610×610×305 mm), 139,3 kg
    18321 l
    16500×3700×305 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7 650,00 €
    7 650,00 €
    1 * 1 ST
       7 650,00 €
  • 119-0363
    Rigid-lock QuickBerm® plus, folded dimensions (610×610×305 mm), 141,1 kg
    Rigid-lock QuickBerm® plus, folded dimensions (610×610×305 mm), 141,1 kg
    22183 l
    17100×4300×305 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8 510,00 €
    8 510,00 €
    1 * 1 ST
       8 510,00 €