Orders LinkContactUsLinkComponent
Personal padlock kit
Supplier:   Brady
Personal padlock kit
Supplier:  Brady
Certificates
Personal padlock kit
Personal padlock kit
3 Options Available Below
Product Details & Documents
This padlock kit includes the equipment needed to implement lockout safety procedures. A convenient way to carry padlocks, hasps and keys in a handy pouch.
  • Pouch has a clear face allowing for easy identification
  • Heavy duty retractable key holder features a Kevlar® cord with split ring attachment and a convertible belt clip/belt loop back attachment
  • Key holder is extendable to 1200 mm
  • Clip can be attached to pouch or be used independently
 : The kit includes: One Safety lockout hasp (25 mm shackle width), one Heavy Duty retractable key holder and two red safety padlocks.
Product Family Options
Product Information
  • Type
    Colour
    Availability
    Price
  • Personal padlock kit, incl. 1× Safety lockout hasp, 1× key holder, 2× safety padlocks
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 109,00 €
    109,00 €
    1 * 1 ST
       109,00 €
Replacement pouch and key holder
Product Information
  • Type
    Colour
    Availability
    Price
  • Personal belt pouch
    Black
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,80 €
    18,80 €
    1 * 1 ST
       18,80 €
  • Heavy duty retractable key holder
    Black
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,10 €
    28,10 €
    1 * 1 ST
       28,10 €
Replacement pouch and key holder
Product Information
  • Catalog #
    Type
    W×D×H
    Colour
    Availability
    Price
  • Safety lockout, 25 mm shackle width
    58,4×0,4×129,3 mm
    Red
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 95,40 €
    95,40 €
    1 * 12 ST
       95,40 €
Product Information
  • Catalog #
    Type
    Availability
    Price
  • Red
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 124,00 €
    124,00 €
    1 * 6 ST
       124,00 €