ContactUsLinkComponent
Avantor® Alltima, HPLC Columns, C8
Avantor® Alltima, HPLC Columns, C8
Supplier:  Avantor
   Sustainable Options Available
Certificates
undefined
Avantor® Alltima, HPLC Columns, C8
Supplier:  Avantor
   Sustainable Options Available
Specifications
  • Catalog No:
  • HICH88372
  • 96081/N
  • HICH88072
  • HICH88073
  • HICH88076
  • HICH88077
  • HICH81392
  • HICH81397
  • HICH81398
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Carbon load:
  • 9%
  • 9%
  • 9%
  • 9%
  • 9%
  • 9%
  • 9%
  • 9%
  • Endcapped:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Surface area:
  • 340 m²/g
  • 340 m²/g
  • 340 m²/g
  • 340 m²/g
  • 340 m²/g
  • 340 m²/g
  • 340 m²/g
  • 340 m²/g
  • USP code:
  • L7
  • L7
  • L7
  • L7
  • L7
  • L7
  • L7
  • L7
  • Environmental certification:
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)