Order Entry
Belgium
ContactUsLinkComponent
Cleaning agent, TICKOPUR RW 77
Cleaning agent, TICKOPUR RW 77
  142-1953
undefined
Cleaning agent, TICKOPUR RW 77
  142-1953

 

  • Capacity:
    2 l
  • Description:
    TICKOPUR RW 77
  • Environmentally Preferable:
  • Pk:
    2 L

 

 

Cleaning agent suitable for cleaning of ultrasonic bath. It can be used to clean glass, metal, ceramics, plastic, rubber and removes grease, oils, wax, protein, pigments, etc.

  • Contains ammonia
  • pH 9,9 at 1%
  • Emulsifying
  • Phosphate-free