Oxytetracycline, Reference Standards
Over dit artikel
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
2 Opties Hieronder Beschikbaar
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
Specifications for Product Family
Specificaties
Specificaties
- Catalog No:
- 1491004.
- 1491015.
- Description:
- Oxytetracycline
- Oxytetracycline Hydrochloride
- CAS No.:
- 6153-64-6
- 2058-46-0
- Formula:
- C₂₂H₂₈N₂O₁₁
- UN No.:
- 3077
Specificaties
Product Family Options
Productinformatie
- DescriptionStandardsPack typePkBeschikbaarheidPrice
- Specificaties:
More
Specifications
Cat. No.1491004.DescriptionOxytetracyclineCAS No.6153-64-6FormulaC₂₂H₂₈N₂O₁₁UN No.3077 - Oxytetracycline HydrochlorideUSP reference standardVial200 mgLog in voor beschikbaarheid1 * 200 mg375,00 €Specificaties:
More
Specifications
Cat. No.1491015.DescriptionOxytetracycline HydrochlorideCAS No.2058-46-0FormulaUN No.



