Order Entry
Belgium
ContactUsLinkComponent
EDTA disodium salt concentrate for dilution to 0.1 mol/l in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
EDTA disodium salt concentrate for dilution to 0.1 mol/l in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4871.
Supplier:  Avantor
CAS Number:  
EDTA disodium salt concentrate for dilution to 0.1 mol/l in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4871.
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Ampoule
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :336.21 g/mol
 :Dose for preparation of 1 litre of solution.
CAS Number:139-33-3
Formula:C₁₀H₁₄N₂Na₂O₈
MDL Number:MFCD00070672