ContactUsLinkComponent
Drums, stackable, with tamper evident cap
Drums, stackable, with tamper evident cap
Catalog # DELT206025
Supplier:  DELTALAB
Drums, stackable, with tamper evident cap
Catalog # DELT206025
Supplier:  DELTALAB
Specifications
  • Capacity:
    25,550 ml
  • Description:
    Drums, with tamper evident cap
  • Pk:
    1
Specifications
About this item
Stackable drums, made of HDPE, natural.
  • Hermetical closure
  • For alimentary use
They embody a handle and a tamper evident, ribbed screw red cap, made of HDPE, with internal joint and internal security ring.