Order Entry
Belgium
Orders LinkContactUsLinkComponent
Alfuzosin, Reference Standards

Alfuzosin, Reference Standards

Supplier:   USP
Alfuzosin, Reference Standards
Supplier:  USP
Certificates

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1012917.
  • 1012928.
  • 1012871.
  • Description:
  • Alfuzosin Hydrochloride
  • Alfuzosin System Suitability Mixture
  • Alfuzosin System Suitability Mixture A
  • Synonym(s):
  • Alfuzosin Hydrochloride containing Impurity A, Impurity D, and Impurity E
  • CAS No.:
  • 81403-68-1
  • Formula:
  • C₁₉H₂₈CLN₅O₄

Specifications

Product Family Options

Product Information

  • Standards
    Pk
    Availability
    Price
  • 1012917.
    Alfuzosin Hydrochloride
    Alfuzosin Hydrochloride
    USP reference standard
    150 mg
    Log in for availability
    Log in for availability
    1 * 150 mg
    1 * 150 mg - 554,00 €
    554,00 €
    1 * 150 mg
       554,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1012917.
    Description
    Alfuzosin Hydrochloride
    Synonym(s)
    CAS No.
    81403-68-1
    Formula
    C₁₉H₂₈CLN₅O₄
  • 1012928.
    Alfuzosin System Suitability Mixture
    Alfuzosin System Suitability Mixture
    USP reference standard
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - 1 350,00 €
    1 350,00 €
    1 * 15 mg
       1 350,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1012928.
    Description
    Alfuzosin System Suitability Mixture
    Synonym(s)
    CAS No.
    Formula
  • 1012871.
    Alfuzosin System Suitability Mixture A
    Alfuzosin System Suitability Mixture A
    USP reference standard
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 350,00 €
    1 350,00 €
    1 * 25 mg
       1 350,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1012871.
    Description
    Alfuzosin System Suitability Mixture A
    Synonym(s)
    Alfuzosin Hydrochloride containing Impurity A, Impurity D, and Impurity E
    CAS No.
    Formula
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.