Order Entry
Belgium
ContactUsLinkComponent
Ranitidine Related Compounds, Reference Standard
Ranitidine Related Compounds, Reference Standard
Supplier:  USP
Certificates
Ranitidine Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • CAS No.:
    91224-69-0
  • Synonym(s):
    5-[[(2-aminoethyl)thio]methyl]-N,N-dimethyl-2-furanmethanamine, hemifumarate salt
  • Formula:
    C₁₄H₂₂N₂O₅S
  • Description:
    Ranitidine Related Compound A

Specifications

Frequently Bought Together