Order Entry
Belgium
Orders LinkContactUsLinkComponent
Pierce™ Spin Columns, Snap Cap

Pierce™ Spin Columns, Snap Cap

Pierce™ spin cups - snap cap
Catalog # PIER69725
Pierce™ Spin Columns, Snap Cap
Catalog # PIER69725
Pierce™ Spin Columns, Snap Cap
Pierce™ Spin Columns, Snap Cap

Pierce™ spin cups - snap cap

Price
219,00 €
Log in for availability
1 * 50 ST
Product Details & Documents

Pierce™ snap cap spin columns are convenient tools for manipulating small volumes of affinity supports (20 to 500 µl) for protein purification.

  • Column volume: 1 ml
  • Resin volume: 20 to 500 µl
  • Filter type: Polyethylene filter, ~30 µm pore size
  • Cap: Attached snap-cap (no cap on collection tube); press-on bottom caps

Applications include affinity purification/affinity chromatography, immunodepletion, immunoprecipitation (IP), and co-immunoprecipitation (co-IP).

Format: Microcentrifuge, spin column
Column type: Empty column

Specifications
  • Description:
    Pierce™ spin cups - snap cap
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.