Volumetric flasks, class A, Duran glass, NS plastic stopper
Supplier: KOEHLER TECHNISCHE PRODUKTEN
Certificates
About this item
Volumetric flasks are made of borosilicate glass with NS plastic (PE) stopper.
- With blue markings
- With standard ground joint
- Good chemical and temperature resistance
Certifications: Class A; DIN 12 664; ISO 1042
Frequently Bought Together