Order Entry
Belgium
Orders LinkContactUsLinkComponent
Acyclovir, Reference Standards

Acyclovir, Reference Standards

Leverancier:   USP
Acyclovir, Reference Standards
Leverancier:  USP
Certificaten

4 Opties Hieronder Beschikbaar

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certificeringen: US Pharmacopea Reference Standard

Specifications for Product Family

Specificaties

Specificaties

  • Catalog No:
  • 1012065.
  • 1012076.
  • 1012054.
  • 1012087.
  • Description:
  • Acyclovir
  • Acyclovir Related Compound A
  • Acyclovir
  • Acyclovir Related Compound G
  • Synonym(s):
  • 2-[(2-amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]ethyl acetate
  • N-[9-[(2-Hydroxyethoxy)methyl]-6-oxo-6,9-dihydro-1H-purin-2-yl]acetamide
  • 2-{[2-(Acetylamino)-6-oxo-1,6-dihydro-9H-purin-9-yl]methoxy}ethyl acetate
  • CAS No.:
  • 59277-89-3
  • 102728-64-3
  • 110104-37-5
  • 75128-73-3
  • Formula:
  • C₈H₁₁N₅O₃
  • C₁₀H₁₃N₅O₄
  • C₁₀H₁₃N₅O₄
  • C₁₂H₁₅N₅O₅

Specificaties

Product Family Options

Productinformatie

  • Description
    Standards
    Pk
    Beschikbaarheid
    Price
  • Acyclovir
    USP reference standard
    300 mg
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 300 mg
    1 * 300 mg - 511,00 €
    511,00 €
    1 * 300 mg
       511,00 €

    Specificaties:
    More
    Specifications
    Cat. No.
    1012065.
    Description
    Acyclovir
    Synonym(s)
    CAS No.
    59277-89-3
    Formula
    C₈H₁₁N₅O₃
  • Acyclovir Related Compound A
    USP reference standard
    50 mg
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 50 mg
    1 * 50 mg - 1 190,00 €
    1 190,00 €
    1 * 50 mg
       1 190,00 €

    Specificaties:
    More
    Specifications
    Cat. No.
    1012076.
    Description
    Acyclovir Related Compound A
    Synonym(s)
    2-[(2-amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]ethyl acetate
    CAS No.
    102728-64-3
    Formula
    C₁₀H₁₃N₅O₄
  • Acyclovir
    USP reference standard
    50 mg
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 50 mg
    1 * 50 mg - 1 110,00 €
    1 110,00 €
    1 * 50 mg
       1 110,00 €

    Specificaties:
    More
    Specifications
    Cat. No.
    1012054.
    Description
    Acyclovir
    Synonym(s)
    N-[9-[(2-Hydroxyethoxy)methyl]-6-oxo-6,9-dihydro-1H-purin-2-yl]acetamide
    CAS No.
    110104-37-5
    Formula
    C₁₀H₁₃N₅O₄
  • Acyclovir Related Compound G
    USP reference standard
    50 mg
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 50 mg
    1 * 50 mg - 1 170,00 €
    1 170,00 €
    1 * 50 mg
       1 170,00 €

    Specificaties:
    More
    Specifications
    Cat. No.
    1012087.
    Description
    Acyclovir Related Compound G
    Synonym(s)
    2-{[2-(Acetylamino)-6-oxo-1,6-dihydro-9H-purin-9-yl]methoxy}ethyl acetate
    CAS No.
    75128-73-3
    Formula
    C₁₂H₁₅N₅O₅
Aanbevelingen

Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.

Anders ontvangt u algemene aanbevelingen.