ContactUsLinkComponent
Quantus® Comfort, Cleanroom Hood
Quantus® Comfort, Cleanroom Hood
Supplier:  VWR Collection
Certificates
undefined
Quantus® Comfort, Cleanroom Hood
Supplier:  VWR Collection
Specifications
  • Catalog No:
  • QNTS031-2CMWH00LID
  • QNTS031-2CMWH0XLID
  • QNTS031-2CMWH00SID
  • QNTS031-2CMWH00MID
  • Garment application:
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Closure type:
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Fabric:
  • 100% polyester weave with conductive yarn
  • 100% polyester weave with conductive yarn
  • 100% polyester weave with conductive yarn
  • 100% polyester weave with conductive yarn
  • Weight:
  • 100,7 g/m²
  • 100,7 g/m²
  • 100,7 g/m²
  • 100,7 g/m²