Order Entry
Belgium
ContactUsLinkComponent
Platinum standard solution
Platinum standard solution
Catalog # PROONIST-3140
CAS Number:  
Platinum standard solution
Catalog # PROONIST-3140
CAS Number:  

Specifications

  • Matrix:
    10% HCl
  • Pack type:
    Ampoule
  • Pk:
    5
  • Element:
    Platinum
  • Symbol:
    Pt

Specifications

About this item

This reference standard is intended for use as a primary calibration standard for the quantitative determination of platinum.

A unit consists of five 10 ml sealed borosilicate glass ampoules of an acidified aqueous solution prepared gravimetrically to contain a known mass fraction of platinum.

Certifications: Traceable to NIST and SRM numbers.