Order Entry
Belgium
Orders LinkContactUsLinkComponent
Calcium standard solution

Calcium standard solution

Calcium, Ca, 10 mg/g, 10% HNO₃, 5
Catalog # PROONIST-3109A
Calcium standard solution
Catalog # PROONIST-3109A
CAS Number:  
Calcium standard solution

Calcium, Ca, 10 mg/g, 10% HNO₃, 5

CAS Number:  
Price
1 730,00 €
Log in for availability
1 * 5 ST
Product Details & Documents

This reference standard solution is intended for use as a primary calibration standard for the quantitative determination of calcium.

A unit consists of five 10 ml sealed borosilicate glass ampoules of an acidified solution prepared gravimetrically to contain a known mass fraction of calcium.

Certifications: Traceable to NIST and SRM numbers.

Specifications

Specifications

  • Concentration:
    10 mg/g
  • Matrix:
    10% HNO₃
  • Pk:
    5
  • Element:
    Calcium
  • Symbol:
    Ca

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.