Order Entry
Belgium
ContactUsLinkComponent
Pyrethroid pesticide and metabolite standards, DCCA
Pyrethroid pesticide and metabolite standards, DCCA
Certificates
Pyrethroid pesticide and metabolite standards, DCCA

Specifications

  • CAS No.:
    55701-05-8
  • Formula:
    C₈H₁₀Cl₂O₂
  • Description:
    trans-DCCA (unlabeled)

Specifications

Frequently Bought Together