Organic reference standards, telmisartans, Ph. Eur., RS
Supplier: EDQM
Certificates
About this item
Specifications
- CAS No.:144701-48-4
- Formula:C₃₃H₃₀N₄O₂
- Description:Telmisartan for peak identification
Specifications
Frequently Bought Together