ContactUsLinkComponent
House dust, standard reference material, NIST-2585
House dust, standard reference material, NIST-2585
Catalog # PROONIST-2585
CAS Number:  
House dust, standard reference material, NIST-2585
Catalog # PROONIST-2585
CAS Number:  
Specifications
  • Pk:
    10 g
  • Description:
    House dust - organic contaminants
Specifications
About this item
This reference standard is intended for use in evaluating analytical methods for the determination of selected of selected polycyclic aromatic hydrocarbons (PAHs), polychlorinated biphenyl (PCB) congers, chlorinated pesticides and polybrominated diphenyl ether (PBDE) cogeners in house dust and similar matrices.
Certifications: Traceable to NIST and SRM.