ContactUsLinkComponent
Baking chocolate standard, NIST-2384
Baking chocolate standard, NIST-2384
Catalog # PROONIST-2384
CAS Number:  
Baking chocolate standard, NIST-2384
Catalog # PROONIST-2384
CAS Number:  
Specifications
  • Pk:
    5
  • Description:
    NIST-2384, baking chocolate
Specifications
About this item
This reference material intended to use in validating methods for determining proximates, fatty acids, calories, vitamins, elements, catechins, caffeine, theobromine, and acrylamide in baking chocolate and similar matrices. It can be used for quality assurance in-house control materials.
A unit consists of five 91 g individually wrapped bars of baking chocolate.
Certifications: Traceable to NIST and SRM.