Order Entry
Belgium
ContactUsLinkComponent
EPA method 1668A/B labelled cleanup standards, in nonane
EPA method 1668A/B labelled cleanup standards, in nonane
Catalog # PROOCIL-EC-4978
CAS Number:  
EPA method 1668A/B labelled cleanup standards, in nonane
Catalog # PROOCIL-EC-4978
CAS Number:  

Specifications

  • Concentration:
    1,2 ml in nonane
  • Pk:
    1,2 mL
  • Description:
    EPA method 1668A /B standard

Specifications

About this item

Standard contains 2,4,4’-TriCB (13C12,99%) 28L - 1000 ng/ml; 2,3,3’,5,5’-PentaCB (13C12,99%) 111L - 1000 ng/ml and
2,2’,3,3’,5,5’,6-HeptaCB (13C12,99%) 178L - 1000 ng/ml.