Order Entry
Belgium
ContactUsLinkComponent
Impurity and API standards, colchicine
Impurity and API standards, colchicine
Certificates
Impurity and API standards, colchicine

Specifications

  • CAS No.:
    6901-13-9
  • Formula:
    C₂₂H₂₅NO₆
  • Description:
    N-[(7S,7bR,10aS)-1,2,3,9-Tetramethoxy-8-oxo-5,6,7,7b,8,10a-hexahydrobenzo[a]cyclopenta[3,4]cyclobuta[1,2-c]cyclohepten-7-yl]acetamide (beta-Lumicolchicine)

Specifications

Frequently Bought Together