Order Entry
Belgium
ContactUsLinkComponent
Impurity and API standard, (2RS)-4-methyl-1-(3-methylpyridin-2-yl)-2-phenylpiperazine
Impurity and API standard, (2RS)-4-methyl-1-(3-methylpyridin-2-yl)-2-phenylpiperazine
Catalog # PROOMM1005.01
CAS Number:  
Impurity and API standard, (2RS)-4-methyl-1-(3-methylpyridin-2-yl)-2-phenylpiperazine
Catalog # PROOMM1005.01
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Standards:
    Reference standards
  • CAS No.:
    191546-94-8
  • Description:
    (2RS)-4-Methyl-1-(3-methylpyridin-2-yl)-2-phenylpiperazine
  • Formula:
    C₁₇H₂₁N₃
  • Description:
    (2RS)-4-Methyl-1-(3-methylpyridin-2-yl)-2-phenylpiperazine

Specifications

About this item