Order Entry
Belgium
ContactUsLinkComponent
Impurity and API standard, 9-hydroxyrisperidone
Impurity and API standard, 9-hydroxyrisperidone
Catalog # PROOMM0491.03
CAS Number:  
Impurity and API standard, 9-hydroxyrisperidone
Catalog # PROOMM0491.03
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Reference standards
  • CAS No.:
    144598-75-4
  • Description:
    (9RS)-3-[2-[4-(6-fluoro-1,2-benzisoxazol-3-yl)piperidin-1-yl]ethyl]-9-hydroxy-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one
  • Formula:
    C₂₃H₂₇FN₄O₃
  • Description:
    (9RS)-3-[2-[4-(6-fluoro-1,2-benzisoxazol-3-yl)piperidin-1-yl]ethyl]-9-hydroxy-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one

Specifications

About this item