Order Entry
Belgium
ContactUsLinkComponent
 
Impurity and API standards, 3-O-methylatorvastatin calcium salt
  PROOMM0400.12
 :  
Impurity and API standards, 3-O-methylatorvastatin calcium salt
  PROOMM0400.12
 :  

 

  • Pk:
    100 mg
  • Standards:
    Reference standards
  • CAS No.:
    887196-29-4
  • Description:
    (3R,5R)-7-[2-(4-Fluorophenyl)-3-phenyl-4-(phenylcarbamoyl)-5-(propan-2-yl)-1H-pyrrol-1-yl]-5-hydroxy-3-methoxyheptanoic acid calcium salt
  • Formula:
    C₆₈H₇₂CaF₂N₄O₁₀
  • Description:
    (3R,5R)-7-[2-(4-Fluorophenyl)-3-phenyl-4-(phenylcarbamoyl)-5-(propan-2-yl)-1H-pyrrol-1-yl]-5-hydroxy-3-methoxyheptanoic acid calcium salt