Impurity and API standards, 2-[[2-[[(4RS)-4-(2-Chlorophenyl)-3-(ethoxycarbonyl)-5-(methoxycarbonyl)-6-methyl-1,4-dihydropyridin-2-yl]methoxy]ethyl]carbamoyl]benzoic acid
PROOMM0383.11
:
- Pk:100 mg
- Standards:Reference standards
- CAS No.:318465-73-5
- Description:2-[[2-[[(4RS)-4-(2-Chlorophenyl)-3-(ethoxycarbonyl)-5-(methoxycarbonyl)-6-methyl-1,4-dihydropyridin-2-yl]methoxy]ethyl]carbamoyl]benzoic acid
- Formula:C₂₈H₂₉ClN₂O₈
- Description:2-[[2-[[(4RS)-4-(2-Chlorophenyl)-3-(ethoxycarbonyl)-5-(methoxycarbonyl)-6-methyl-1,4-dihydropyridin-2-yl]methoxy]ethyl]carbamoyl]benzoic acid